2-Amino-5-chlorchinazolin

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Chloroquinazolin-2-amine

OR110444-10G OR110444-1G OR110444-5G
APOSOR110444-10GEA 282 EUR
APOSOR110444-10G APOSOR110444-1G APOSOR110444-5G
2-Amino-5-chlorchinazolin
2-Amino-5-chloroquinazoline
Formel: C₈H₆ClN₃
Molecular Weight: 179,606 g/mol
Schmelzpunkt: 249…252 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00974335
CAS-Nummer: 190273-70-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR