3-(Dimethylamino)-1-(2-thienyl)-2-buten-1-one

Lieferant: Apollo Scientific
Achtung

OR110455-1G OR110455-5G
APOSOR110455-1GEA 89.9 EUR
APOSOR110455-1G APOSOR110455-5G
3-(Dimethylamino)-1-(2-thienyl)-2-buten-1-one
3-(Dimethylamino)-1-(2-thienyl)-2-buten-1-one
Formel: C₁₀H₁₃NOS
Molecular Weight: 195,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03791167
CAS-Nummer: 112677-16-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR