7-Bromo-5-methylbenzo[b]furan

Lieferant: Apollo Scientific

Synonyme: 7-Bromo-5-methylbenzofuran

OR110462-1G OR110462-250MG OR110462-5G
APOSOR110462-1GEA 183 EUR
APOSOR110462-1G APOSOR110462-250MG APOSOR110462-5G
7-Bromo-5-methylbenzo[b]furan
7-Bromo-5-methylbenzo[b]furan
Formel: C₉H₇BrO
Molecular Weight: 211.058 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11877828
CAS-Nummer: 35700-48-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR