3-Bromo-5-chloro-2-hydroxybenzyl alcohol

Lieferant: Apollo Scientific
Achtung

OR110463-1G OR110463-250MG OR110463-5G
APOSOR110463-1GEA 98.6 EUR
APOSOR110463-1G APOSOR110463-250MG APOSOR110463-5G
3-Bromo-5-chloro-2-hydroxybenzyl alcohol
3-Bromo-5-chloro-2-hydroxybenzyl alcohol
Formel: C₇H₆BrClO₂
Molecular Weight: 237,48 g/mol
Schmelzpunkt: 64…66 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06202648
CAS-Nummer: 116795-47-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR