2-(4-Methylphenoxy)benzenecarbaldehyde

Lieferant: Apollo Scientific

Synonyme: 2-(4-Methylphenoxy)benzaldehyde

OR110497-1G OR110497-5G
APOSOR110497-1GEA 125 EUR
APOSOR110497-1G APOSOR110497-5G
2-(4-Methylphenoxy)benzenecarbaldehyde
2-(4-Methylphenoxy)benzenecarbaldehyde
Formel: C₁₄H₁₂O₂
Molecular Weight: 212,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01568738
CAS-Nummer: 19434-35-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR