5-Chloro-3-phenyl[1,2,4]thiadiazole

Lieferant: Apollo Scientific

OR110572-1G OR110572-5G
APOSOR110572-1GEA 112 EUR
APOSOR110572-1G APOSOR110572-5G
5-Chloro-3-phenyl[1,2,4]thiadiazole
5-Chloro-3-phenyl[1,2,4]thiadiazole
Formel: C₈H₅ClN₂S
Molecular Weight: 196,66 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00096450
CAS-Nummer: 24255-23-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR