6-Chloro-2-oxo-2H-chromene-3-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR110616-1G
OR110616-5G
APOSOR110616-1GEA
43.5
EUR
APOSOR110616-1G
APOSOR110616-5G
6-Chloro-2-oxo-2H-chromene-3-carboxylic acid
6-Chloro-2-oxo-2H-chromene-3-carboxylic acid
Formel:
C₁₀H₅ClO₄ Molecular Weight: 224.6 g/mol Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00991129 CAS-Nummer: 883-92-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...