4,5,6,7-Tetrahydro-1-benzothiophene-2-carbohydrazide

Lieferant: Apollo Scientific

OR110625-1G OR110625-5G OR110625-10G
APOSOR110625-1GEA 186 EUR
APOSOR110625-1G APOSOR110625-5G APOSOR110625-10G
4,5,6,7-Tetrahydro-1-benzothiophene-2-carbohydrazide
4,5,6,7-Tetrahydro-1-benzothiophene-2-carbohydrazide
Formel: C₉H₁₂N₂OS
Lagertemperatur: Raumtemperatur
MDL: MFCD04969092
CAS-Nummer: 65361-27-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR