2-(3-Chlorophenyl)-1,3-thiazole-4-carbonyl chloride

Lieferant: Apollo Scientific

OR110682-1G OR110682-5G
APOSOR110682-1GEA 125 EUR
APOSOR110682-1G APOSOR110682-5G
2-(3-Chlorophenyl)-1,3-thiazole-4-carbonyl chloride
2-(3-Chlorophenyl)-1,3-thiazole-4-carbonyl chloride
Formel: C₁₀H₅Cl₂NOS
Molecular Weight: 258.13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08060545
CAS-Nummer: 868755-69-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR