5-Chlor-1,3-dimethyl-1H-pyrazol-4-sulfonsäure
Lieferant: Apollo Scientific
OR110683-1G
OR110683-5G
OR110683-10G
APOSOR110683-1GEA
125
EUR
APOSOR110683-1G
APOSOR110683-5G
APOSOR110683-10G
5-Chlor-1,3-dimethyl-1H-pyrazol-4-sulfonsäure
5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulphonic acid
Formel:
C₅H₇ClN₂O₃S Lagertemperatur: Raumtemperatur |
CAS-Nummer:
499785-48-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...