4-Chloro-6-ethoxy-2-pyrimidinamine

Lieferant: Apollo Scientific
Achtung

OR110708-1G OR110708-5G
APOSOR110708-1GEA 43.5 EUR
APOSOR110708-1G APOSOR110708-5G
4-Chloro-6-ethoxy-2-pyrimidinamine
4-Chloro-6-ethoxy-2-pyrimidinamine
Formel: C₆H₈ClN₃O
Molecular Weight: 173,6000 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00457060
CAS-Nummer: 89784-02-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR