Ethylimidazo[1,2-a]pyridin-6-carboxylat 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR110709-1G
OR110709-5G
APOSOR110709-1GEA
77
EUR
APOSOR110709-1G
APOSOR110709-5G
Ethylimidazo[1,2-a]pyridin-6-carboxylat 95%
Ethylimidazo[1,2-a]pyridin-6-carboxylat
Formel:
C₁₀H₁₀N₂O₂ Molecular Weight: 190,20 g/mol Schmelzpunkt: 97…99 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD13176746 CAS-Nummer: 158001-04-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...