2-(Chlormethyl)-6-nitrobenzoxazol
Lieferant: Apollo Scientific
|
Achtung
|
OR110716-1G
OR110716-250MG
OR110716-5G
APOSOR110716-1GEA
267
EUR
APOSOR110716-1G
APOSOR110716-250MG
APOSOR110716-5G
2-(Chlormethyl)-6-nitrobenzoxazol
2-(Chloromethyl)-6-nitrobenzoxazole
Formel:
C₈H₅ClN₂O₃ Schmelzpunkt: 69…70 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00775454 CAS-Nummer: 221638-74-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...