6-Nitro-2H-1,4-benzoxazin-3(4H)-one 98%
Lieferant: Apollo Scientific
Synonyme:
6-Nitro-2H-benzo[b][1,4]oxazin-3(4H)-one
OR110726-1G
OR110726-5G
OR110726-25G
APOSOR110726-1GEA
30.6
EUR
APOSOR110726-1G
APOSOR110726-5G
APOSOR110726-25G
6-Nitro-2H-1,4-benzoxazin-3(4H)-one 98%
6-Nitro-2H-1,4-benzoxazin-3(4H)-one
Formel:
C₈H₆N₂O₄ Molecular Weight: 194.15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03425744 CAS-Nummer: 81721-87-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...