6-Amino-2H-1,4-benzoxazin-3(4H)-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Amino-3,4-dihydro-3-oxo-2H-1,4-benzoxazine,6-Amino-3-oxo-3,4-dihydrobenzo[1,4]oxazine,6-Amino-2H-benzo[b][1,4]oxazin-3(4H)-one,6-Amino-4H-benzo[1,4]oxazin-3-one, 6-amino-2,4-dihydro-1,4-benzoxazin-3-one

OR110727-1G OR110727-5G
APOSOR110727-1GEA 65.4 EUR
APOSOR110727-1G APOSOR110727-5G
6-Amino-2H-1,4-benzoxazin-3(4H)-one
6-Amino-2H-1,4-benzoxazin-3(4H)-one
Formel: C₈H₈N₂O₂
Molecular Weight: 164.17 g/mol
Schmelzpunkt: 266…268 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03425742
CAS-Nummer: 89976-75-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR