Methyl-4-amino-2-morpholino-1,3-thiazole-5-carboxylate

Lieferant: Apollo Scientific
Achtung

OR110730-1G OR110730-250MG
APOSOR110730-1GEA 285 EUR
APOSOR110730-1G APOSOR110730-250MG
Methyl-4-amino-2-morpholino-1,3-thiazole-5-carboxylate
Methyl-4-amino-2-morpholino-1,3-thiazole-5-carboxylate
Formel: C₉H₁₃N₃O₃S
Molecular Weight: 243,29 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00139418
CAS-Nummer: 99967-78-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR