7-Nitro-2H-1,4-benzoxazin-3(4H)-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Nitro-2H-benzo[b][1,4]oxazin-3(4H)-one

OR110745-1G OR110745-5G OR110745-25G
APOSOR110745-1GEA 26.1 EUR
APOSOR110745-1G APOSOR110745-5G APOSOR110745-25G
7-Nitro-2H-1,4-benzoxazin-3(4H)-one
7-Nitro-2H-1,4-benzoxazin-3(4H)-one
Formel: C₈H₆N₂O₄
Molecular Weight: 194.15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00090355
CAS-Nummer: 81721-86-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR