3-Bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR110746-1G OR110746-5G OR110746-250MG
APOSOR110746-1GEA 190 EUR
APOSOR110746-1G APOSOR110746-5G APOSOR110746-250MG
3-Bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylic acid
3-Bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylic acid
Formel: C₈H₄BrClN₂O₂
Schmelzpunkt: 216…219 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08064717
CAS-Nummer: 923225-00-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR