3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Oxo-3,4-dihydro-2H-benzo[b][1,4]oxazine-7-carboxylic acid
OR110758-1G
OR110758-250MG
APOSOR110758-1GEA
168
EUR
APOSOR110758-1G
APOSOR110758-250MG
3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylic acid 95%
3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylic acid
Formel:
C₉H₇NO₄ Molecular Weight: 193,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07774185 CAS-Nummer: 214848-62-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...