5-Amino-7-brom-1H-indazol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Bromo-1H-indazol-5-amine

OR110843-1G OR110843-250MG
APOSOR110843-1GEA 29 EUR
APOSOR110843-1G APOSOR110843-250MG
5-Amino-7-brom-1H-indazol 98%
5-Amino-7-bromo-1H-indazole
Formel: C₇H₆BrN₃
Molecular Weight: 212,0500 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08275114
CAS-Nummer: 953411-10-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR