Methyl-1-butyl-5-oxopyrrolidine-3-carboxylate

Lieferant: Apollo Scientific

OR110898-1G OR110898-5G
APOSOR110898-1GEA 232 EUR
APOSOR110898-1G APOSOR110898-5G
Methyl-1-butyl-5-oxopyrrolidine-3-carboxylate
Methyl-1-butyl-5-oxopyrrolidine-3-carboxylate
Formel: C₁₀H₁₇NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD15143238
CAS-Nummer: 59857-87-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR