1-(3-Amino-4-hydroxyphenyl)ethanone
Lieferant: Apollo Scientific
|
Gefahr
|
OR110942-1G
OR110942-250MG
APOSOR110942-1GEA
186
EUR
APOSOR110942-1G
APOSOR110942-250MG
1-(3-Amino-4-hydroxyphenyl)ethanone
1-(3-Amino-4-hydroxyphenyl)ethanone
Formel:
C₈H₉NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06657507 CAS-Nummer: 54255-50-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...