Ethyl-(4-iodphenyl)acetat

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2-(4-iodophenyl)acetate

OR110943-1G OR110943-25G OR110943-5G
APOSOR110943-1GEA 21.9 EUR
APOSOR110943-1G APOSOR110943-25G APOSOR110943-5G
Ethyl-(4-iodphenyl)acetat
Ethyl-(4-iodphenyl)acetat
Formel: C₁₀H₁₁IO₂
Molecular Weight: 290,0976 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07779004
CAS-Nummer: 15250-46-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR