1H-Indole-2-carboxamide
Lieferant: Apollo Scientific
|
Achtung
|
OR110958-1G
OR110958-250MG
OR110958-5G
APOSOR110958-1GEA
115
EUR
APOSOR110958-1G
APOSOR110958-250MG
APOSOR110958-5G
1H-Indole-2-carboxamide
1H-Indole-2-carboxamide
Formel:
C₉H₈N₂O Schmelzpunkt: 225…227 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00231036 CAS-Nummer: 1670-84-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...