2-(4-Bromo-1H-pyrazol-1-yl)pyrimidine
Lieferant: Apollo Scientific
|
Achtung
|
OR110960-1G
OR110960-250MG
APOSOR110960-1GEA
178
EUR
APOSOR110960-1G
APOSOR110960-250MG
2-(4-Bromo-1H-pyrazol-1-yl)pyrimidine
2-(4-Bromo-1H-pyrazol-1-yl)pyrimidine
Formel:
C₇H₅BrN₄ Molecular Weight: 225,05 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09834926 CAS-Nummer: 857641-46-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...