1-Methyl-1H-indole-3-carboxylic acid

Lieferant: Apollo Scientific

Synonyme: 1-Methylindole-3-carboxylic acid

OR111128-10G OR111128-1G OR111128-5G
APOSOR111128-10GEA 68.3 EUR
APOSOR111128-10G APOSOR111128-1G APOSOR111128-5G
1-Methyl-1H-indole-3-carboxylic acid
1-Methyl-1H-indole-3-carboxylic acid
Formel: C₁₀H₉NO₂
Molecular Weight: 175,184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01321244
CAS-Nummer: 32387-21-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR