N,N-Diisopropyl-4-nitrobenzamide

Lieferant: Apollo Scientific

OR111133-1G OR111133-5G
APOSOR111133-1GEA 0 EUR
APOSOR111133-1G APOSOR111133-5G
N,N-Diisopropyl-4-nitrobenzamide
N,N-Diisopropyl-4-nitrobenzamide
Formel: C₁₃H₁₈N₂O₃
Lagertemperatur: Raumtemperatur
CAS-Nummer: 79606-48-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR