2-Piperidinonicotinonitril

Lieferant: Apollo Scientific

OR111151-1G OR111151-5G
APOSOR111151-1GEA 0 EUR
APOSOR111151-1G APOSOR111151-5G
2-Piperidinonicotinonitril
2-Piperidinonicotinonitril
Formel: C₁₁H₁₃N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD04113613
CAS-Nummer: 34595-23-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR