2-Pyrrolidin-1-ylnicotinonitrile
Lieferant: Apollo Scientific
OR111153-10G
OR111153-1G
OR111153-5G
APOSOR111153-10GEA
0
EUR
APOSOR111153-10G
APOSOR111153-1G
APOSOR111153-5G
2-Pyrrolidin-1-ylnicotinonitrile
2-Pyrrolidin-1-ylnicotinonitrile
Formel:
C₁₀H₁₁N₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD07792921 CAS-Nummer: 59025-38-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...