(6-Bromoimidazo[1,2-a]pyridin-2-yl)acetic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(6-Bromoimidazo[1,2-a]pyridin-2-yl)acetic acid

OR111221-1G OR111221-5G OR111221-10G
APOSOR111221-1GEA 37.7 EUR
APOSOR111221-1G APOSOR111221-5G APOSOR111221-10G
(6-Bromoimidazo[1,2-a]pyridin-2-yl)acetic acid
(6-Bromoimidazo[1,2-a]pyridin-2-yl)acetic acid
Formel: C₉H₇BrN₂O₂
Molecular Weight: 255.08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06739269
CAS-Nummer: 59128-15-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR