Ethyl-2,5-dichloronicotinate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 2,5-dichloronicotinate
OR111287-1G
OR111287-5G
APOSOR111287-1GEA
105
EUR
APOSOR111287-1G
APOSOR111287-5G
Ethyl-2,5-dichloronicotinate
Ethyl-2,5-dichloronicotinate
Formel:
C₈H₇Cl₂NO₂ Molecular Weight: 220,06 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07636850 CAS-Nummer: 148065-10-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...