Dimethyl-2-(2-chloro-6-nitrophenyl)malonate
Lieferant: Apollo Scientific
OR111301-10G
OR111301-5G
APOSOR111301-10GEA
0
EUR
APOSOR111301-10G
APOSOR111301-5G
Dimethyl-2-(2-chloro-6-nitrophenyl)malonate
Dimethyl-2-(2-chloro-6-nitrophenyl)malonate
Formel:
C₁₁H₁₀ClNO₆ Molecular Weight: 287.66 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03787565 CAS-Nummer: 147124-36-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...