6-Chloropyridine-2-carbonyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
OR111351-1G
APOSOR111351-1GEA
235
EUR
APOSOR111351-1G
6-Chloropyridine-2-carbonyl chloride
6-Chloropyridine-2-carbonyl chloride
Formel:
C₆H₃Cl₂NO Lagertemperatur: Kühlschrank |
MDL:
MFCD00274531 CAS-Nummer: 80099-98-5 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...