5-Amino-1,3-dimethyl-1H-pyrazole-4-carbonitrile
Lieferant: Apollo Scientific
OR111442-1G
OR111442-5G
APOSOR111442-1GEA
0
EUR
APOSOR111442-1G
APOSOR111442-5G
5-Amino-1,3-dimethyl-1H-pyrazole-4-carbonitrile
5-Amino-1,3-dimethyl-1H-pyrazole-4-carbonitrile
Formel:
C₆H₈N₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00128277 CAS-Nummer: 54820-92-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...