6-Bromo-7-methylquinoxaline
Lieferant: Apollo Scientific
|
Achtung
|
OR111498-1G
OR111498-250MG
APOSOR111498-1GEA
944
EUR
APOSOR111498-1G
APOSOR111498-250MG
6-Bromo-7-methylquinoxaline
6-Bromo-7-methylquinoxaline
Formel:
C₉H₇BrN₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD16987756 CAS-Nummer: 646504-80-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...