2-Bromonaphthalen-1-ol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromo-1-naphthalenol
OR111504-1G
APOSOR111504-1GEA
21.9
EUR
APOSOR111504-1G
2-Bromonaphthalen-1-ol
2-Bromonaphthalen-1-ol
Formel:
C₁₀H₇BrO Molecular Weight: 223,07 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00274158 CAS-Nummer: 771-15-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...