3-Chloro-1H-indazol-5-amine

Lieferant: Apollo Scientific

OR111527-1G OR111527-250MG
APOSOR111527-1GEA 589 EUR
APOSOR111527-1G APOSOR111527-250MG
3-Chloro-1H-indazol-5-amine
3-Chloro-1H-indazol-5-amine
Formel: C₇H₆ClN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD07781654
CAS-Nummer: 41330-49-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR