Ethyl-3-amino-2,6-dichloroisonicotinate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 3-amino-2,6-dichloropyridine-4-carboxylate, Ethyl 3-amino-2,6-dichloroisonicotinate
OR111558-1G
OR111558-5G
APOSOR111558-1GEA
39.3
EUR
APOSOR111558-1G
APOSOR111558-5G
Ethyl-3-amino-2,6-dichloroisonicotinate
Ethyl-3-amino-2,6-dichloroisonicotinate
Formel:
C₈H₈Cl₂N₂O₂ Molecular Weight: 235.07 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16661134 CAS-Nummer: 458543-81-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...