2,5-Dihydroxy-p-benzochinon

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,5-Dihydroxy-1,4-benzoquinone

OR11157-25G OR11157-5G
APOSOR11157-25GEA 58 EUR
APOSOR11157-25G APOSOR11157-5G
2,5-Dihydroxy-p-benzochinon
2,5-Dihydroxy-p-benzochinon
Formel: C₆H₄O₄
Molecular Weight: 140,1 g/mol
Schmelzpunkt: 220 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001598
CAS-Nummer: 615-94-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR