5,6-Dimethoxybenzimidazol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5,6-Dimethoxy-1H-benzo[d]imidazole

OR11164-1G OR11164-250MG OR11164-5G
APOSOR11164-1GEA 88.6 EUR
APOSOR11164-1G APOSOR11164-250MG APOSOR11164-5G
5,6-Dimethoxybenzimidazol 97%
5,6-Dimethoxybenzimidazole
Formel: C₉H₁₀N₂O₂
Molecular Weight: 178,19 g/mol
Schmelzpunkt: 179…185 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00612461
CAS-Nummer: 72721-02-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR