2-Quinoxalinol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Quinoxalinone, 2(1H)-Quinoxalinone, 2-Hydroxyquinoxaline, quinoxalin-2-ol

OR11198-500G
APOSOR11198-500GEA 82.8 EUR
APOSOR11198-500G
2-Quinoxalinol 98%
2-Quinoxalinol
Formel: C₈H₆N₂O
Molecular Weight: 146,15 g/mol
Schmelzpunkt: 271…272 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006722
CAS-Nummer: 1196-57-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR