6-methoxyquinoxaline

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Methoxyquinoxaline

OR11203-1G
APOSOR11203-1GEA 75.4 EUR
APOSOR11203-1G
6-methoxyquinoxaline
6-methoxyquinoxaline
Formel: C₉H₈N₂O
Molecular Weight: 160,176 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00666601
CAS-Nummer: 6639-82-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR