3,5-Dichloro-4-pyridone-1-acetic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
[3,5-Dichloro-4-oxopyridin-1(4H)-yl]acetic acid
OR11204-250G
OR11204-50G
APOSOR11204-250GEA
66.7
EUR
APOSOR11204-250G
APOSOR11204-50G
3,5-Dichloro-4-pyridone-1-acetic acid
3,5-Dichloro-4-pyridone-1-acetic acid
Formel:
C₇H₅Cl₂NO₃ Molecular Weight: 222.03 g/mol Schmelzpunkt: 239…241 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02180547 CAS-Nummer: 56187-37-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...