4-Iodcumol 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Iodisopropylbenzol, 2-(4-Iodphenyl)propan
OR11205-25G
OR11205-5G
APOSOR11205-25GEA
0
EUR
APOSOR11205-25G
APOSOR11205-5G
4-Iodcumol 95%
4-Iodcumol
Formel:
C₉H₁₁I Molecular Weight: 246,09 g/mol Siedepunkt: 236…238 °C Dichte: 1,525 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013711 CAS-Nummer: 17356-09-1 EINECS: 241-384-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...