2-Nitroterephthalsäure-1-methylester 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Methyl 2-nitroterephthalate
OR11224-25G
OR11224-5G
APOSOR11224-25GEA
0
EUR
APOSOR11224-25G
APOSOR11224-5G
2-Nitroterephthalsäure-1-methylester 95%
2-Nitroterephthalsäure-1-methylester
Formel:
C₉H₇NO₆ Molecular Weight: 225,17 g/mol Schmelzpunkt: 176…178 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00024510 CAS-Nummer: 35092-89-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...