3'-Nitropropiophenon

Lieferant: Apollo Scientific
Achtung

Synonyme: m-Nitropropiophenon

OR11232-5G
APOSOR11232-5GEA 116 EUR
APOSOR11232-5G
3'-Nitropropiophenon
3'-Nitropropiophenon
Formel: C₉H₉NO₃
Molecular Weight: 179,18 g/mol
Schmelzpunkt: 99…102 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051516
CAS-Nummer: 17408-16-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR