Phenethyl isocyanate 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Isocyanic Acid Phenethyl Ester, (2-Isocyanatoethyl)benzene, Isocyanic Acid 2-Phenylethyl Ester, 2-Phenylethyl Isocyanate

OR11236-25G OR11236-5G OR11236-100G
APOSOR11236-25GEA 72.5 EUR
APOSOR11236-25G APOSOR11236-5G APOSOR11236-100G
Phenethyl isocyanate 98%
Phenethyl isocyanate

  • May form small amount of sediment.

Formel: C₉H₉NO
Molecular Weight: 147.18 g/mol
Siedepunkt: 210 °C
Dichte: 1.063 g/cm³ (25 °C)
Flammpunkt: 100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00037044
CAS-Nummer: 1943-82-4
EINECS: 413-080-0
UN: 2922
ADR: 8+6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR