5-Cyan-2-thiophenboronsäure 97%
Lieferant: Apollo Scientific
|
Gefahr
|
APOSOR11252-10GEA
179
EUR
APOSOR11252-10G
APOSOR11252-1G
APOSOR11252-5G
5-Cyan-2-thiophenboronsäure 97%
5-Cyano-2-thiopheneboronic acid
Formel:
C₅H₄BNO₂S Molecular Weight: 152,97 g/mol Schmelzpunkt: 125…128 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02094029 CAS-Nummer: 305832-67-1 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...