N-(9H-Purin-6-yl)benzamide 94%

Lieferant: Apollo Scientific
Achtung

OR11259-25G OR11259-5G
APOSOR11259-25GEA 30.6 EUR
APOSOR11259-25G APOSOR11259-5G
N-(9H-Purin-6-yl)benzamide 94%
N-(9H-Purin-6-yl)benzamide
Formel: C₁₂H₉N₅O
Molecular Weight: 239,24 g/mol
Schmelzpunkt: 241…244 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00443632
CAS-Nummer: 4005-49-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR