5-Brom-2'-chlor-2-hydroxybenzophenon
Lieferant: Apollo Scientific
|
Achtung
|
OR1128-10G
OR1128-1G
OR1128-5G
APOSOR1128-10GEA
259
EUR
APOSOR1128-10G
APOSOR1128-1G
APOSOR1128-5G
5-Brom-2'-chlor-2-hydroxybenzophenon
5-Brom-2'-chlor-2-hydroxybenzophenon
Formel:
C₁₃H₈BrClO₂ Schmelzpunkt: 104…108 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02682999 CAS-Nummer: 332104-54-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...